Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PPLLIQDEDS TRIERVMDNN TTVKMVPIKI VHSESQPEKE SRQSLACPAE PPALPHGLEK DQIKTLSTSE QFYS
Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.
The protein encoded by this gene shares significant similarities with the apical protein from Xenopus laevis which is implicated in amiloride-sensitive sodium channel activity. This gene is a strong candidate gene for ocular albinism type 1 syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apical-like protein; APX homolog of Xenopus; Protein APXL; Protein Shroom2
Gene Aliases: APXL; HSAPXL; SHROOM2
UniProt ID: (Human) Q13796
Entrez Gene ID: (Human) 357
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support