Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: DGESIDTYER DLYLSVQFNL NQTTLTSTIL KRGYINKSET RLGSLHVWGK GTTPVNAVTL TYNGNKNSLP FNEDTTNMIL RIDLTTHNVT LEEPIEINW
This gene encodes a sucrase-isomaltase enzyme that is expressed in the intestinal brush border. The encoded protein is synthesized as a precursor protein that is cleaved by pancreatic proteases into two enzymatic subunits sucrase and isomaltase. These two subunits heterodimerize to form the sucrose-isomaltase complex. This complex is essential for the digestion of dietary carbohydrates including starch, sucrose and isomaltose. Mutations in this gene are the cause of congenital sucrase-isomaltase deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-glucosidase; oligosaccharide alpha-1,6-glucosidase; sucrase-isomaltase (alpha-glucosidase); Sucrase-isomaltase, intestinal
Gene Aliases: SI
UniProt ID: (Human) P14410
Entrez Gene ID: (Human) 6476
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support