Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EVNFPLLLNC FGQPGTKWIP FSYTYRRPLR THYGYINVKT QEPLQLDCDL
Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.
Involved in the biosynthesis of ganglioside GD1A from GM1B. Transfers CMP-NeuAc with an alpha-2,6-linkage to GalNAc residue on NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc of glycoproteins and glycolipids. ST6GalNAcIII prefers glycolipids to glycoproteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 3; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase III; GalNAc alpha-2,6-sialyltransferase III; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) C; Sialyltransferase 7C; SIAT7-C; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltran; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 3; ST6 GalNAc alpha-2,6-sialyltransferase 3; ST6GALNAC III; ST6GalNAcIII; STY
Gene Aliases: PRO7177; SIAT7C; ST6GALNAC3; ST6GALNACIII; STY; UNQ2787/PRO7177
UniProt ID: (Human) Q8NDV1
Entrez Gene ID: (Human) 256435
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support