Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MSRQTATALP TGTSKCPPSQ RVPALTGTTA SNNDLASLFE CPVCFDYVLP PILQCQSGHL VCSNCRPKLT CCPTCRGPLG SIRNLAMEKV ANSVLFPCKY ASSGCEITLP
This gene encodes a protein that is a member of the seven in absentia homolog family. The protein is an E3 ligase and is involved in ubiquitination and proteasome-mediated degradation of specific proteins. The activity of this ubiquitin ligase has been implicated in the development of certain forms of Parkinson's disease, the regulation of the cellular response to hypoxia and induction of apoptosis. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase SIAH1; FLJ08065; h HUMSIAH; hSIAH1; RING-type E3 ubiquitin transferase SIAH1; Seven in absentia homolog 1; Siah-1; Siah-1a
Gene Aliases: HUMSIAH; SIAH1; SIAH1A
UniProt ID: (Human) Q8IUQ4
Entrez Gene ID: (Human) 6477
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support