Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 6
Sequence Similarities: 100% Mouse/Rat.
Tissue Specificity: Widely expressed at a low level. Down-regulated in advanced hepatocellular carcinomas.
Positive Control: HCT 116 cell lysate, RAW264.7 cell lysate, mouse liver tissue lysate, mouse brain tissue lysate, rat liver tissue lysate, rat brain tissue lysate, Rat testis testis lysate, A549.
Subcellular Location: Cytoplasm, Nucleus.
Predicted band size: 31 kDa.
This gene encodes a protein that is a member of the seven in absentia homolog family. The protein is an E3 ligase and is involved in ubiquitination and proteasome-mediated degradation of specific proteins. The activity of this ubiquitin ligase has been implicated in the development of certain forms of Parkinson's disease, the regulation of the cellular response to hypoxia and induction of apoptosis. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase SIAH1; E3 ubiquitin-protein ligase SIAH1A; E3 ubiquitin-protein ligase SIAH1B; FLJ08065; h HUMSIAH; hSIAH1; mSiah-1a; RING-type E3 ubiquitin transferase SIAH1; RING-type E3 ubiquitin transferase SIAH1A; RING-type E3 ubiquitin transferase SIAH1B; seven in absentia 1A; Seven in absentia homolog 1; Seven in absentia homolog 1a; Seven in absentia homolog 1b; Siah-1; Siah-1a; Siah-1b
Gene Aliases: AA960570; AA982064; AI853500; HUMSIAH; SIAH; siah-1b; SIAH1; SIAH1A; Siah1b; Sinh1a; Sinh1b
UniProt ID: (Human) Q8IUQ4, (Rat) Q920M9, (Mouse) P61092, (Mouse) Q06985
Entrez Gene ID: (Human) 6477, (Rat) 140941, (Mouse) 20437, (Mouse) 20438
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support