Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Mouse anti Human Siglec-10 antibody, clone 5G6 recognizes human Siglec-10 (Sialic acid-binding Ig-like lectin 10), a putative adhesion molecule and member of the Ig superfamily, expressed by monocytes, B cells, eosinophils, and at a higher level by a subpopulation of CD16+CD56- natural killer (NK) cells.
SIGLECs are members of the immunoglobulin superfamily that are expressed on the cell surface. Most SIGLECs have 1 or more cytoplasmic immune receptor tyrosine-based inhibitory motifs, or ITIMs. SIGLECs are typically expressed on cells of the innate immune system, with the exception of the B-cell expressed SIGLEC6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: sialic acid binding Ig-like lectin 10 Ig-like lectin 7; Sialic acid-binding Ig-like lectin 10; Siglec; Siglec-10; siglec-like gene 2; Siglec-like protein 2
Gene Aliases: PRO940; SIGLEC-10; SIGLEC10; SLG2; UNQ477/PRO940
UniProt ID: (Human) Q96LC7
Entrez Gene ID: (Human) 89790
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support