Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: EGDRQYGDGY LLQVQELVTV QEGLCVHVPC SFSYPQDGWT DSDPVHGYWF RAGDRPYQDA PVATNNPDRE VQAETQGRFQ LLGDIWSNDC SLSIRDARKR DKGS
SIGLEC8 is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It preferentially binds to alpha2,3-linked sialic acid. and also binds to alpha2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. SIGLEC8 is expressed specifically on eosinophils. The protein contains 1 copy of a cytoplasmic motif that is referred to as the immunoreceptor tyrosine-based inhibitor motif (ITIM). This motif is involved in downmodulation of cellular responses. The phosphorylated ITIM motif binds to the SH2 domain of PTPN6/SHP-1. The SIGLEC8 gene belongs to the immunoglobulin superfamily.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDw329; MGC59785; SAF-2; sialic acid binding Ig-like lectin 8; Sialic acid-binding Ig-like lectin 8; Sialoadhesin family member 2; Siglec; Siglec-8
Gene Aliases: SAF2; SIGLEC-8; SIGLEC8; SIGLEC8L
UniProt ID: (Human) Q9NYZ4
Entrez Gene ID: (Human) 27181
Molecular Function: immunoglobulin superfamily cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support