Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western blots, approximately 20% cross-reactivity with recombinant mouse SIGNR1 is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD209; CD209 antigen-like protein E; DC-SIGN-related protein 4; DC-SIGNR4
Gene Aliases: Cd209e; mSIGNR4; SIGNR4
UniProt ID: (Mouse) Q91ZW7
Entrez Gene ID: (Mouse) 170780
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support