Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKIQQLHPAN AKTVEWDWKL LTYVMEEEGQ TLPGRVLFLR YVVQTLEDDF QQTLRRQRQH LQQSIANMVL SCDKQPHNVR DVIKWLVKA
Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
SIMC1 (SUMO Interacting Motifs Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includeSUMO polymer binding. An important paralog of this gene isSKIDA1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ44216; oocyte maturation associated 1; Platform element for inhibition of autolytic degradation; SUMO-interacting motif-containing protein 1; SUMO-interacting motifs containing 1
Gene Aliases: C5orf25; OOMA1; PLEIAD; SIMC1
UniProt ID: (Human) Q8NDZ2
Entrez Gene ID: (Human) 375484
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support