Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 17
Keep as concentrated solution.
Predicted reactivity: Mouse (96%), Rat (94%), Chicken (86%), Rhesus Monkey (100%).
Positive Control: 293T, A431, HeLa, HepG2, mouse brain, rat brain.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Mammalian Sin3 (mSin3) is a 150 kDa protein closely related to the yeast SIN3 repressor protein involved in the transcriptional repression of many genes. There are two mammalian isoforms, mSin3A and mSin3B. Containing 4 paired amphipathic helix domains (PAH domains), mSin3A and mSin3B have been shown to directly interact with several other transcriptional repressor proteins including HDAC1, HDAC2, RbAp46, the methyl CpG binding protein MeCP2, the Mad/Max heterodimer, and the corepressors silencing mediator of retinoic acid and thyroid hormone receptor (SMRT) and nuclear receptor corepressor (N-CoR).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Histone deacetylase complex subunit Sin3a; Mammalian Sin3A; Paired amphipathic helix protein Sin3a; SIN; SIN3 homolog A, transcription regulator; SIN3 transcription regulator homolog A; SIN3A; transcriptional co-repressor Sin3A; Transcriptional corepressor Sin3a; transcriptional regulator, SIN3 yeast homolog A; transcriptional regulator, SIN3A
Gene Aliases: AW553200; Kiaa4126; mKIAA4126; mSin3A; Sin3; SIN3A
UniProt ID: (Human) Q96ST3, (Mouse) Q60520
Entrez Gene ID: (Human) 25942, (Rat) 363067, (Mouse) 20466
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support