Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: HKMVFIVNSE DYMYRRGTLC RAKQVQPLVL LRHHQHFEEW HSRWLEDNVT VEAASLVQDW LMGEEDEDMV PCKTLCETVH VHGLPVTRYR VQYSRRPA
Acts as a transcriptional repressor. Interacts with MXI1 to repress MYC responsive genes and antagonize MYC oncogenic activities. Interacts with MAD-MAX heterodimers by binding to MAD. The heterodimer then represses transcription by tethering SIN3B to DNA. Also forms a complex with FOXK1 which represses transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Histone deacetylase complex subunit Sin3b; Paired amphipathic helix protein Sin3b; SIN3 homolog B, transcriptional regulator; SIN3 transcription regulator homolog B; SIN3B; Transcriptional corepressor Sin3b
Gene Aliases: KIAA0700; SIN3B
UniProt ID: (Human) O75182
Entrez Gene ID: (Human) 23309
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support