Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GQPIPESGD PKGTPKSDAE PEPGNLSEKV SHLESMLRKL QEDLQKEKAD RAALEEEVRS LRHNNRRLQA ESESAATRLL LASKQLGSPT ADLA (950-1042 aa encoded by BC010492)
The product of this gene is a mitogen induced GTPase activating protein. It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localized to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterized to date.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTPase-activating protein Spa-1; MGC102688; MGC17037; p130 SPA-1; signal-induced proliferation-associated gene 1; Signal-induced proliferation-associated protein 1; Sipa-1
Gene Aliases: SIPA1; SPA1
UniProt ID: (Human) Q96FS4
Entrez Gene ID: (Human) 6494
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support