Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen Sequence: SSSMADFRKF FAKAKHIVII SGAGVSAESG VPTFRGAGGY WRKWQAQDLA TPLAFAHNPS RVWEFYHYRR EVMGSKEPNA GHRAIAECET RLGKQGRRVV VITQNIDELH RKAGTKNLLE IHGSLFKTRC TSCGVVAENY KSPICPALSG KGAPEPGTQD ASIPVEKLPR CEEAGCGGLL RPHVVWFGEN LDPAILEEVD RELAHCDLCL VVGTSSVVYP AAMFAPQVAA RGVPVAEFNT ETTPATNRFR FHFQGPCGTT LPEALACHEN ETVS
SIRT5 is a human member of a family of proteins called Sirtuins (Sir2-like proteins) and are present in prokaryotes and eukaryotes. All Sir2-like proteins have a sirtuin core domain, which contains a series of sequence motifs conserved in organisms ranging from bacteria to humans. Bacterial, yeast and mammalian sirtuins are able to metabolize NAD and possibly at as mono-ADP-ribosyltransferases. The enzymatic function of sirtuins is not yet completely understood but recent reports of histone-activated Sir2-mediated NAD metabolism and NAD-activated Sir2-mediated histone deacetylation suggest a possible coupled reciprocal activation mechanism involving interactions of Sir2 with NAD and the N epsilon-acetyl-lysine groups of acetylated histones.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ36950; NAD-dependent deacetylase sirtuin-5; NAD-dependent lysine demalonylase and desuccinylase sirtuin-5, mitochondrial; NAD-dependent protein deacylase sirtuin-5, mitochondrial; NAD-dependent protein deacylase sirtuin-5, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03160}; Regulatory protein SIR2 homolog 5; regulatory protein SIR2 homolog 5 {ECO:0000255|HAMAP-Rule:MF_03160}; silent mating type information regulation 2, S.cerevisiae, homolog 5; sir2-like 5; SIR2-like protein 5; SIR2-like protein 5 {ECO:0000255|HAMAP-Rule:MF_03160}; SIRT5; sirtuin type 5
Gene Aliases: 0610012J09Rik; 1500032M05Rik; AV001953; SIR2L5; SIRT5
UniProt ID: (Human) F5H5Z9, (Mouse) Q8K2C6, (Rat) Q68FX9
Entrez Gene ID: (Human) 23408, (Mouse) 68346, (Rat) 306840
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support