Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Sirtuins regulate epigenetic gene silencing. SIRT6 (Mono-ADP-ribosyltransferase sirtuin-6), a mouse Sir2 homolog, is a chromatin-associated nuclear ADP-ribosyltransferase protein. It appears to be involved in DNA repair and may also play a role in human aging.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSIRT6; mSIRT6; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; NAD-dependent protein deacylase sirtuin-6; Protein mono-ADP-ribosyltransferase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; SIR2-like protein 6; sir2-related protein type 6; sirtuin type 6
Gene Aliases: 2810449N18Rik; AI043036; SIR2L6; SIRT6
UniProt ID: (Human) Q8N6T7, (Mouse) P59941
Entrez Gene ID: (Human) 51548, (Mouse) 50721
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support