Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: GQDLLSVPMT QAALGEIVPT AEDQVGHPSP AVHQDFVQEH RLVLQSVANM KENFLSNSES KATSSLMMLD SKSKYVLDGM VDTVCEDLET DKKELAKLQT VQLDEDMQD
SIX4 gene ontology annotations related to this gene include anatomical structure development; embryonic cranial skeleton morphogenesis; fungiform papilla morphogenesis; generation of neurons; inner ear morphogenesis; male gonad development; male sex determination; metanephric mesenchyme development; myoblast migration; myotome development; negative regulation of neuron apoptotic process; negative regulation of satellite cell differentiation; negative regulation of transcription, DNA-templated; olfactory placode formation; pharyngeal system development; positive regulation of branching involved in ureteric bud morphogenesis; positive regulation of ureteric bud formation; protein localization to nucleus; regulation of branch elongation involved in ureteric bud branching; regulation of epithelial cell proliferation; regulation of protein localization; regulation of synaptic growth at neuromuscular junction; sarcomere organization; skeletal muscle fiber differentiation; thymus development; trigeminal ganglion development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein SIX4; Sine oculis homeobox homolog 4
Gene Aliases: AREC3; SIX4
UniProt ID: (Human) Q9UIU6
Entrez Gene ID: (Human) 51804
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support