Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Neutrophil and pancreatic elastase-specific inhibitor of skin. It may prevent elastase-mediated tissue proteolysis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Elafin; Elastase-specific inhibitor; ESI; Peptidase inhibitor 3; peptidase inhibitor 3, skin-derived; PI-3; pre-elafin; protease inhibitor 3, skin-derived (SKALP); Protease inhibitor WAP3; SKALP; Skin-derived antileukoproteinase; trappin-2; WAP four-disulfide core domain 14; WAP four-disulfide core domain protein 14
Gene Aliases: cementoin; ESI; PI3; SKALP; WAP3; WFDC14
UniProt ID: (Human) P19957
Entrez Gene ID: (Human) 5266
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support