Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPNPSSTSS PYPLPEEIRN LLADVETFVA DILKGENLSK KAKEKRESLI KKIKDVKSIY LQEFQDKGDA EDGEEYDDPF AGPPDTISLA SERYDKDDEA PSDGAQFPPI AAQDLPFVLK AGYLEKRRKD HSFLGFEWQK RWCALSKTVF YYYGSDKDKQ QKGEFAIDGY SVRMNNTLRK DGKKDCCFEI SAPDKRIYQF TAASPKDAEE WVQQLKFVLQ DMESDIIPED YDERGELYDD VDHPLPISNP LTSSQPIDDE IYEELPEEEE DSAPVKVEEQ RKMSQDSVHH TSGDKSTDYA NFYQGLWDCT GAFSDELSFK RGDVIYILSK EYNRYGWWVG EMKGAIGLVP KAYIMEMYDI (1-359 aa encoded by BC036044)
Fyb (Fyn binding protein) and the anchoring proteins SKAP55 and SKAP55-R (SKAP55-related protein) associate with the tyrosine kinase p59fyn. SKAP55 and SKAP55-R bind to Fyb through their SH3 domains and function as substrates for p59Fyn in resting T cells. SKAP55 contains an N-terminal pleckstrin homology domain and a C-terminal SH3 domain binding motif of adjacent arginine and lysine residues followed by tandem tyrosines (i.e. RKxxYxxY). SKAP55-R, similar in overall structure to SKAP55, contains a coiled-coil N-terminal domain. SKAP55 associates with SLAP-130, another component of the Fyn complex, which plays a role in the regulation of signaling events initiated by lymphocyte antigen receptors leading up to T cell activation. The human SKAP55 gene maps to chromosome 17q21.32 and encodes a 359 amino acid protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fyn-associated phosphoprotein SKAP55 homologue; MGC10411; MGC33304; Pyk2/RAFTK-associated protein; Retinoic acid-induced protein 70; SKAP-55HOM; SKAP-HOM; SKAP55 homolog; SKAP55 homologue; SKAP55-HOM; src family associated phosphoprotein 2; Src family-associated phosphoprotein 2; Src kinase-associated phosphoprotein 2; Src kinase-associated phosphoprotein 55-related protein; src kinase-associated phosphoprotein of 55-related protein; Src-associated adapter protein with PH and SH3 domains; Src-associated adaptor protein
Gene Aliases: 2610021A10Rik; AA960083; BB137539; mSKAP55R; PRAP; RA70; SAPS; SCAP2; Scap55r; SKAP-HOM; SKAP2; SKAP55R
UniProt ID: (Human) O75563, (Rat) Q920G0, (Mouse) Q3UND0
Entrez Gene ID: (Human) 8935, (Pig) 100625903, (Rat) 155183, (Mouse) 54353
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support