Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
Mouse anti Pig SLA Class II DR antibody, clone 289/13 recognizes SLA DR molecules which are expressed on all B cells, antigen presenting cells and on certain subsets of resting and activated T cells.
SLA Class II DR recognizes SLA DR molecules which are expressed on all B cells, antigen presenting cells and on certain subsets of resting and activated T cells. The major histocompatibility complex (MHC) is a cluster of genes that are important in the immune response to infections. In pigs, this is referred to as the swine leukocyte antigen (SLA) region. There are 3 major MHC class II proteins encoded by the SLA which are SLA DP, SLA DQ and SLA DR.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: MHC class II antigen; MHC class II SLA-DRB1-5; SLA-DR-beta class II light chain; SLA-DR1 beta chain; SLA-DRB*K04G; SLA-DRB*K05C(A); SLA-DRB*K05G; SLA-DRB*K06G; SLA-DRB*K07G; SLA-DRB*K08G; SLA-DRB*K09G
Gene Aliases: DRB1; LA-DRB-c; LA-DRB-d; SLA-DR-alpha; SLA-DRA1; SLA-DRA2; SLA-DRB; SLADRB
Entrez Gene ID: (Pig) 106504372, (Pig) 100135040, (Pig) 100153386
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support