Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs, no cross-reactivity with recombinant human (rh) SLAMF7 or rhSLAMF6 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a member of the CD2 family of cell surface proteins involved in lymphocyte activation. These proteins are characterized by Ig domains. This protein is expressed in lymphoid tissues, and studies of a similar protein in mouse suggest that it may function during B cell lineage commitment. The gene is found in a region of chromosome 1 containing many CD2 genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B lymphocyte activator macrophage expressed; B-lymphocyte activator macrophage expressed; BCM-like membrane protein; CD353; FLJ20442; MGC129578; SLAM family member 8
Gene Aliases: BLAME; CD353; SBBI42; SLAMF8
UniProt ID: (Human) Q9P0V8
Entrez Gene ID: (Human) 56833
Molecular Function: immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support