Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: ALPLLEMPEE KDLRSSNEDS HIVKIEKLNE RSKRKDDGVA HRDSAGQRCI CLSKAVGYLT GDMKEYRIWL KDKHLALQFI DWVLRGTAQ
UT-A1, a kidney-specific urea transporter is expressed in the renal collecting duct where it mediates trans-epithelial urea transport and is a target for regulation by vasopressin. Urea movement out of the collecting duct in the inner medulla of the kidney allows accumulation of urea in the medullary interstitium, thereby allowing maximum water reabsorption from the collecting ducts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16167; hUT-A6; HUT2; MGC119566; MGC119567; solute carrier family 14 (urea transporter), member 2; Solute carrier family 14 member 2; Urea transporter 2; Urea transporter, kidney; urea transporter-2; UT1; UTA; UTA1; UTR
Gene Aliases: hUT-A6; HUT2; SLC14A2; UT-A2; UT2; UTA; UTR
UniProt ID: (Human) Q15849
Entrez Gene ID: (Human) 8170
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support