Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MATKTELSPT ARESKNAQDM QVDETLIPRK VPSLCSARYG IALVLHFCNF TTIAQNVIMN ITMVAMVNST SPQSQLNDSS EVLPVDSFGG LSKAPKSLPT KSSILGGQFA IWEKWGPPQE RSRLCSIALS GMLLGCFTAI LIGGFISETL GWPFVFYIFG GVGCVCCLLW FVVIYDDPVS YPWISTSEKE YIISSLKQQV GSSKQPLPIK AMLRSLPIWS ICLGCFSHQW LVSTMVVYIP TYISSVYHVN IRDNGLLSAL PFIVAWVIGM VGGYLADFLL TKKFRLITVR KIATILGSLP SSALIVSLPY LNSGYITATA LLTLSCGLST LCQSGIYINV LDIAPRYSSF LMGASRGFSS IAPVIVPTVS GFLLSQDPEF GWRNVFFLLF AVNLLGLLFY LIFGEADVQE WAKERKLTRL
SLC17A3 may be involved in actively transporting phosphate into cells via Na(+) cotransport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Na(+)/PI cotransporter 4; NPT4; Sodium-dependent phosphate transport protein 4; Sodium/phosphate cotransporter 4; solute carrier family 17 (organic anion transporter), member 3; solute carrier family 17 (sodium phosphate), member 3; Solute carrier family 17 member 3
Gene Aliases: GOUT4; NPT4; SLC17A3; UAQTL4
UniProt ID: (Human) O00476
Entrez Gene ID: (Human) 10786
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support