Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTLTSRRQD SQEARPECQA WTGTLLLGTC LLYCARSSMP ICTVSMSQDF GWNKKEAGIV LSSFFWGYCL TQVVGGHLGD RIGGEK (1-85 aa encoded by BC025312)
Involved in vesicular storage and exocytosis of ATP. May accumulate ATP and other nucleotides in secretory vesicles such as adrenal chromaffin granules and synaptic vesicles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: solute carrier family 17 (vesicular nucleotide transporter), member 9; Solute carrier family 17 member 9; Vesicular nucleotide transporter; vesicular nucleotide transporter SLC17A9; VNUT; Voltage-gated purine nucleotide uniporter SLC17A9
Gene Aliases: 1700019H03Rik; BC019537; C20orf59; POROK8; SLC17A9; VNUT
UniProt ID: (Human) Q9BYT1, (Mouse) Q8VCL5
Entrez Gene ID: (Human) 63910, (Mouse) 228993
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support