Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PIVPTFLYD MEFKEVNSSL HLGHAGSSPH ALASPAFSTI FSFFNNNTVA VEESVPSGIA WMNDTASTIP PPATEAISAH KNNCLQGTGF LEEEITR (43-138 aa encoded by BC009387)
SLC18A1 (Solute Carrier Family 18 Member A1) is a Protein Coding gene. Diseases associated with SLC18A1 include Barbiturate Abuse and Adrenal Neuroblastoma. Among its related pathways are Transport of inorganic cations/anions and amino acids/oligopeptides and Synaptic vesicle pathway. Gene Ontology (GO) annotations related to this gene include monoamine transmembrane transporter activity and xenobiotic transmembrane transporter activity. An important paralog of this gene is SLC18A2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CGAT; Chromaffin granule amine transporter; solute carrier family 18 (vesicular monoamine transporter), member 1; solute carrier family 18 (vesicular monoamine), member 1; Solute carrier family 18 member 1; solute carrier family 18, member 1; solute carrier family18 (vesicular monoamine) member 1 (chromaffin granule amine transporter); VAT1; Vesicular amine transporter 1; vesicular monoamine transporter 1; VMAT; VMAT1
Gene Aliases: CGAT; SLC18A1; VAT1; VMAT-1; VMAT1
UniProt ID: (Human) P54219, (Rat) Q01818
Entrez Gene ID: (Human) 6570, (Rat) 25693
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support