Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: PMPQKSLFFH HIPSTCQRVN GIKVQNGGIV TDTPASNHLP GWEDIESKIP LNMEEPPVEE PEPKPDRLLV LKVLWND
SLC19A2 encodes a thiamin transporter protein. Mutations in this gene cause thiamin-responsive megaloblastic anemia syndrome (TRMA), which is an autosomal recessive disorder characterized by diabetes mellitus, megaloblastic anemia and sensorineural deafness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: high affinity thiamine transporter; reduced folate carrier protein (RFC) like; solute carrier family 19 (thiamine transporter), member 2; Solute carrier family 19 member 2; TC1; Thiamine carrier 1; Thiamine transporter 1; ThTr-1
Gene Aliases: SLC19A2; TC1; THMD1; THT1; THTR1; TRMA
UniProt ID: (Human) Q9UKJ2
Entrez Gene ID: (Human) 10560
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support