Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTKSNGEEPK MGGRMERFQQ GVSKRTLLAK KKVQNITKED VKSYLFRNAF VLLTVTAVIV GTILGFTLRP YRMSYREVKY FSFPGELLMR MLQMLVLPLI ISSLVTGMAA LDSKASGKMG MRAVVYYMTT TIIAVVIGII IVIIIHPGKG TKENMHREGK IVRVTAADAF LDLIRNMFPP NLVEACFKQF KTNYEKRSFK VPIQANETLV GAVINNVSEA METLTRITEE LVPVPGSVNG VNALGLVVFS MCFGFVIGNM KEQGQALREF FDSLNEAIMR LVAVIMWYAP VGILFLIAGK IVEMEDMGVI GGQLAMYTVT VIVGLLIHAV IVLPLLYFLV TRKNPWVFIG GLLQALITAL GTSSSSATLP ITFKCLEENN GVDKRVTRFV LPVGATINMD GTALYEALAA IFIAQVNNFE LNFGQIITIS ITATAASIGA AGIPQAGLVT MVIVLTSVGL PTDDITLIIA VDWFLDRLRT TTNVLGDSLG AGIVEHLSRH ELKNRDVEMG NSVIEENEMK KPYQLIAQDN ETEKPIDSET KM
This gene encodes a member of a member of a high affinity glutamate transporter family. Mutations in this gene are associated with episodic ataxia, Type 6. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EAAT-1; Excitatory amino acid transporter 1; FLJ25094; GLAST-1; Sodium-dependent glutamate/aspartate transporter 1; solute carrier family 1 (glial high affinity glutamate transporter), member 3; Solute carrier family 1 member 3
Gene Aliases: EA6; EAAT1; GLAST; GLAST1; SLC1A3
UniProt ID: (Human) P43003
Entrez Gene ID: (Human) 6507
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support