Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.
The antigen is homologous in rat.
Specificity of this antibody: EAAT1.
This gene encodes a member of a member of a high affinity glutamate transporter family. Mutations in this gene are associated with episodic ataxia, Type 6. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EAAT-1; Excitatory amino acid transporter 1; FLJ25094; GLAST; GLAST-1; Glial glutamate transporter; Glial high affinity glutamate transporter; GluT-1; glutamate transporter; glutamate/aspartate transporter; High-affinity neuronal glutamate transporter; Sodium-dependent glutamate/aspartate transporter 1; Solute carrier family 1 member 3; solute carrier family 1, member 3
Gene Aliases: AI504299; B430115D02Rik; EAAT1; GLAST; GLAST-1; GLU-T; GluT-1; Gmt1; MGluT1; Slc1a3
UniProt ID: (Rat) P24942, (Mouse) P56564
Entrez Gene ID: (Rat) 29483, (Mouse) 20512
Molecular Function: primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support