Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total URAT1.
SLC22A12 is required for efficient urate re-absorption in the kidney. SLC22A12 regulates blood urate levels. SLC22A12 mediates saturable urate uptake by facilitating the exchange of urate against organic anions.The protein encoded by this gene is a urate transporter and urate-anion exchanger which regulates the level of urate in the blood. This protein is an integral membrane protein primarily found in kidney. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Organic anion transporter 4-like protein; Renal-specific transporter; RST; solute carrier family 22 (organic anion/cation transporter), member 12; solute carrier family 22 (organic anion/urate transporter), member 12; solute carrier family 22 (organic cation transporter)-like 2; Solute carrier family 22 member 12; URAT1; Urate anion exchanger 1; urate transporter 1; Urate:anion antiporter SLC22A12
Gene Aliases: AI987855; OAT4L; OATL4; RST; SLC22A12; Slc22al2; UNQ6453/PRO34004; URAT1
UniProt ID: (Human) Q96S37, (Rat) Q3ZAV1, (Mouse) Q8CFZ5
Entrez Gene ID: (Human) 116085, (Rat) 365398, (Mouse) 20521
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support