Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RVGGIIAPF IPSLKYVQWS LPFIVFGATG LTSGLLSLLL PETLNSPLLE TFSDLQVYSY RRLGEEALSL QALDPQQCVD KESSLGSESE EEEEFYDADE ETQMIK (443-547 aa encoded by BC026358)
Organic ion transporters, such as SLC22A15, transport various medically and physiologically important compounds, including pharmaceuticals, toxins, hormones, neurotransmitters, and cellular metabolites. These transporters are also referred to as amphiphilic solute facilitators (ASFs).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Flipt 1; fly-like putative organic ion transporter 1; Fly-like putative transporter 1; solute carrier family 22 (organic cation transporter), member 15; Solute carrier family 22 member 15; solute carrier family 22, member 15; trans-like protein
Gene Aliases: 2610034P21Rik; 4930477M19; A530052I06Rik; BB219042; FLIPT1; PRO34686; SLC22A15; UNQ9429/PRO34686
UniProt ID: (Human) Q8IZD6, (Mouse) Q504N2
Entrez Gene ID: (Human) 55356, (Mouse) 242126
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support