Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSALGRSSV ILLTYVLAAT ELTCLFMQFS IVPYLSRKLG LDSIAFGYLQ TTFGVLQLLG GPVFGRFADQ RGARAALTLS FLAALALYLL LAAASSPALP GVYLLFASRL PGALMHTLPA AQMVITDLSA PEERPAALGR LGLCFGVGVI LGSLLGGTLV SAYGIQCPAI LAALATLLGA VLSFTCIPAS TKGAKTDAQA PLPGGPRASV FDLKAIASLL RLPDVPRIFL VKVASNCPTG LFMVMFSIIS MDFFQLEAAQ AGYLMSFFGL LQMVTQGLVI GQLSSHFSEE VLLRASVLVF IVVGLAMAWM SSVFHFCLLV PGLVFSLCTL NVVTDSMLIK AVSTSDTGTM LGLCASVQPL LRTLGPTVGG LLYRSFGVPV FGHVQVAINT LVLLVLWRKP MPQRKDKVR (17-424 aa encoded by BC015571)
ORCTL2 (organic cation transporter-like protein 2), also known as HET, ITM, BWR1A, IMPT1, TSSC5, BWSCR1A, SLC22A1L, p45-BWR1A or SLC22A18, is a 424 amino acid multi-pass membrane protein that localizes at the apical membrane surface of renal proximal tubules. ORCTL2 is expressed at high levels in the kidney, liver, colon and in fetal renal proximal tubules and present at lower levels in heart, brain and lung. Belonging to the major facilitator superfamily, ORCTL2 may act as a transporter of organic cations based on a proton efflux antiport mechanism and may also play a role in the transport of chloroquine and quinidine-related compounds in kidney. Defects in ORCTL2 are associated with breast and lung cancer and are the cause of rhabdomyosarcoma type 1, a malignant tumor derived from striated muscle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene A protein; Efflux transporter-like protein; imprinted multi-membrane spanning polyspecific transporter-related protein 1; Imprinted multi-membrane-spanning polyspecific transporter-related protein 1; Multi-membrane-spanning polyspecific transporter; ORCTL-2; Organic cation transporter-like protein 2; organic cationic transporter-like 2; p45 Beckwith-Wiedemann region 1A; p45-Beckwith-Wiedemann region 1 A; p45-BWR1A; solute carrier family 22 (organic cation transporter), member 18; Solute carrier family 22 member 1-like; Solute carrier family 22 member 18; solute carrier family 22, member 18; tumor suppressing subtransferable candidate 5; Tumor-suppressing STF cDNA 5 protein; Tumor-suppressing subchromosomal transferable fragment candidate gene 5 protein
Gene Aliases: AW260131; BWR1A; BWSCR1A; HET; IMPT1; ITM; ORCTL2; p45-BWR1A; SLC22A18; SLC22A1L; TSSC5
UniProt ID: (Human) Q96BI1, (Mouse) Q78KK3, (Rat) Q6AY78
Entrez Gene ID: (Human) 5002, (Mouse) 18400, (Rat) 309131
Molecular Function: transporter secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support