Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TMIRQTGMGM GSTMARVGSI VSPLVSMTAE LYPSMPLFIY GAVPVAASAV TVLLPETLGQ PLPDTVQDLE SRKGKQTRQQ QEHQKYMVPL QASAQEKNGL
The protein is involved in the sodium-dependent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and may be localized to the basolateral membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hOAT1; hPAHT; hROAT1; Organic anion transporter 1; PAH transporter; para-aminohippurate transporter; Renal organic anion transporter 1; solute carrier family 22 (organic anion transporter), member 6; Solute carrier family 22 member 6
Gene Aliases: HOAT1; OAT1; PAHT; ROAT1; SLC22A6
UniProt ID: (Human) Q4U2R8
Entrez Gene ID: (Human) 9356
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support