Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: TPESIRWLVL SGKSSKALKI LRRVAVFNGK KEEGERLSLE ELKLNLQKEI SLAKAKYTAS DLFRIPMLRR
SLC22A8 is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. SLC22A8 is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney.The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hOAT3; MGC24086; Oat3; Organic anion transporter 3; Organic anion/dicarboxylate exchanger; rOat3; solute carrier family 22 (organic anion transporter), member 8; solute carrier family 22 ,member 8; Solute carrier family 22 member 8
Gene Aliases: OAT3; OCT3; Roct; SLC22A8
UniProt ID: (Human) Q8TCC7, (Rat) Q9R1U7
Entrez Gene ID: (Human) 9376, (Rat) 83500
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support