Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAATASAGAG GIDGKPRTSP KSVKFLFGGL AGMGATVFVQ PLDLVKNRMQ LSGEGAKTRE YKTSFHALTS ILKAEGLRGI YTGLSAGLLR QATYTTTRLG IYTVLFERLT GADGTPPGFL LKAVIGMTAG ATGAFVGTPA EVALIRMTAD GRLPADQRRG YKNVFNALIR ITREEGVLTL WRGCIPTMAR AVVVNAAQLA SYSQSKQFLL DSGYFSDNIL CHFCASMISG LVTTAASMPV DIAKTRIQNM RMIDGKPEYK NGLDVLFKVV RYEGFFSLWK GFTPYYARLG PHTVLTFIFL EQMNKAYKRL FLSG
The oxoglutarate/malate carrier transports 2-oxoglutarate across the inner membranes of mitochondria in an electroneutral exchange for malate or other dicarboxylic acids (summary by Iacobazzi et al., 1992 [PubMed 1457818]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial 2-oxoglutarate/malate carrier protein; OGCP; SLC25A11; solute carrier family 20 (oxoglutarate carrier), member 4; solute carrier family 25 (mitochondrial carrier; oxoglutarate carrier), member 11; Solute carrier family 25 member 11
Gene Aliases: OGC; SLC20A4; SLC25A11
UniProt ID: (Human) Q02978
Entrez Gene ID: (Human) 8402
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support