Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: Mouse liver, Mouse kidney
Immunogen sequence: IDAFHQIIRD EGISALWNGT FPSLLLVFNP AIQFMFYEGL KRQLLKKRMK LSSLDVFIIG AVAKAIATTV TYPLQTVQSI LRFGRHRLNP ENRTLGSLRN I
This protein encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Studies suggest that this protein may regulate cytokine production, and thus play a key role in the control of the inflammatory response.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 34 kDa peroxisomal membrane protein; peroxisomal integral membrane protein 47; Peroxisomal membrane protein PMP34; peroxisomal membrane protein, 34 kDa; solute carrier family 25 (mitochondrial carrier; peroxisomal membrane protein, 34kDa), member 17; Solute carrier family 25 member 17; solute carrier family 25, member 17
Gene Aliases: 34kDa; 47kDa; PMP34; Pmp35; Pmp47; SLC25A17
UniProt ID: (Human) O43808, (Mouse) O70579
Entrez Gene ID: (Human) 10478, (Mouse) 20524
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support