Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ELILQSIDV DGTMTVDWNE WRDYFLFNPV TDIEEIIRFW KHSTGIDIGD SLTIPDEFTE DEKKSGQWWR QLLAGGIAGA VSRTSTAPLD RLKIMMQVHG SKSDKMNIFG GFRQMVKEGG IRSLWRGNGT NVIKIAPETA VKFWAYEQYK KLLTEEGQKI GTFERFISGS MAGATAQTFI YPMEVMKTRL AVGKTGQYSG IYDCAKKILK HEGLGAFYKG YVPNLLGIIP YAGIDLAVYE LLKSYWLDNF AKDSVNPGVM VLLGCGALSS TCGQLASYPL ALVRTRMQAQ AMLEGSPQLN MVGLFRRIIS KEGIPGLYRG ITPNFMKVLP AVGISYVVYE NMKQTLGVTQ K (128-477 aa encoded by BC068561)
SLC25A24 is a calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. It may act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium-binding mitochondrial carrier protein SCaMC-1; calcium-binding transporter; DKFZp586G0123; Mitochondrial adenyl nucleotide antiporter SLC25A24; Mitochondrial ATP-Mg/Pi carrier protein 1; Mitochondrial Ca(2+)-dependent solute carrier protein 1; RP11-356N1.3; SCaMC-1; short calcium-binding mitochondrial carrier 1; Short calcium-binding mitochondrial carrier protein 1; Small calcium-binding mitochondrial carrier protein 1; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 24; Solute carrier family 25 member 24
Gene Aliases: 2610016M12Rik; APC1; MCSC1; SCAMC-1; SCAMC1; SLC25A24
UniProt ID: (Human) Q6NUK1, (Mouse) Q8BMD8
Entrez Gene ID: (Human) 29957, (Mouse) 229731, (Rat) 310791
Molecular Function: mitochondrial carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support