Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Mitochondrial carriers, including SLC25A26, are a family of transport proteins found mostly in the inner membranes of mitochondria. They shuttle metabolites and cofactors through the mitochondrial membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial S-adenosylmethionine carrier protein; mitochondrial S-adenosylmethionine transporter; S-adenosylmethionine mitochondrial carrier protein; SAM carrier; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 26; solute carrier family 25 (S-adenosylmethionine carrier), member 26; Solute carrier family 25 member 26
Gene Aliases: 4930433D19Rik; 4933433F13Rik; AW557176; COXPD28; D6Bwg0781e; SAMC; SLC25A26; Slc25a6
UniProt ID: (Human) Q70HW3, (Mouse) Q5U680
Entrez Gene ID: (Human) 115286, (Mouse) 67582
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support