Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGLTFINAL VFGVQGNTLR ALGHDSPLNQ FLAGAAAGAI QCVICCPMEL AKTRLQLQDA GPARTYKGSL DCLAQIYGHE GLRGVNRGMV STLLRETPSF GVYFLTYDAL TRALGCEPGD RLLVPKLLLA GGTSGIVSWL STYPVDV (1-146 aa encoded by BC132966)
SLC25A29 belongs to the mitochondrial carrier family and it may has palmitoylcarnitine transporting activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CACT-like; carnitine-acylcarnitine translocase like; Carnitine/acylcarnitine translocase-like; Mitochondrial basic amino acids transporter; Mitochondrial carnitine/acylcarnitine carrier protein CACL; Mitochondrial ornithine transporter 3; solute carrier family 25 (mitochondrial carnitine/acylcarnitine carrier), member 29; solute carrier family 25 (mitochondrial carrier, palmitoylcarnitine transporter), member 29; Solute carrier family 25 member 29; solute carrier family 25, member 29
Gene Aliases: C030003J19Rik; C14orf69; CACL; mCACL; ORNT3; SLC25A29
UniProt ID: (Human) Q8N8R3, (Mouse) Q8BL03, (Rat) Q5HZE0
Entrez Gene ID: (Human) 123096, (Mouse) 214663, (Rat) 314441
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support