Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
SLC25A37 belongs to the SLC25 family of mitochondrial carrier proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial iron transporter 1; Mitochondrial solute carrier protein; mitoferrin; Mitoferrin-1; predicted protein of HQ2217; solute carrier family 25 (mitochondrial iron transporter), member 37; Solute carrier family 25 member 37; solute carrier family 25, member 37
Gene Aliases: HT015; MFRN; MFRN1; MSC; MSCP; PRO1278; PRO1584; PRO2217; SLC25A37
UniProt ID: (Human) Q9NYZ2
Entrez Gene ID: (Human) 51312
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support