Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
SLC25A42 belongs to the SLC25 family of mitochondrial carrier proteins.The exact function of SLC25A42 remains unknown.SLC25A42 belongs to the SLC25 family of mitochondrial carrier proteins (Haitina et al., 2006 [PubMed 16949250]).[supplied by OMIM]. Sequence Note: removed 2 bases from the 5' end that did not align to the reference genome assembly. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-3160 BC045598.1 3-3162
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial coenzyme A transporter SLC25A42; Solute carrier family 25 member 42
Gene Aliases: SLC25A42
UniProt ID: (Human) Q86VD7
Entrez Gene ID: (Human) 284439
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support