Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SLC25A46 belongs to the SLC25 family of mitochondrial carrier proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial outer membrane protein SLC25A46; Solute carrier family 25 member 46; solute carrier family 25, member 46
Gene Aliases: 1200007B05Rik; AI325987; HMSN6B; SLC25A46; TB1
UniProt ID: (Human) Q96AG3, (Mouse) Q9CQS4
Entrez Gene ID: (Human) 91137, (Mouse) 67453
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support