Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MTDAAVSFAK DFLAGGVAAA ISKTAVAPIE RVKLLLQVQH ASKQITADKQ YKGIIDCVVR IPKEQGVLSF WRGNLANVIR YFPTQALNFA FKDKYKQIFL GGVDKRTQFW RYFAGNLASG GAAGATSLCF VYPLDFARTR LAADVGKAGA EREFRGLGDC LVKIYKSDGI KGLYQGFNVS VQGIIIYRAA YFGIYDTAKG MLPDPKNTHI VISWMIAQTV TAVAGLTSYP FDTVRRRMMM QSGRKGTDIM YTGTLDCWRK IARDEGGKAF FKGAWSNVLR GMGGAFVLVL YDEIKKYT
Adenine nucleotide translocator (ANT) and the voltage-dependent anion-selective channel proteins 1 and 2 (VDAC1 and VDAC2) are components of the permeability transition pore complex (PTPC) of the mitochondrial inner and outer membranes, respectively. Formation of PTPCs, the subsequent dissipation of mitochondrial inner membrane potential and release of cytochrome c through the outer mitochondrial membrane are critical events in the early stages of apoptosis. Bax, a proapoptotic protein, has been shown to act upon ANT to induce the dissipation of mitochondrial inner membrane potential. ANT1 has a role in the maintenance of mitochondrial DNA by catalyzing the exchange of ADP and ATP across the mitochondrial inner membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenine nucleotide translocator 2; adenine nucleotide translocator 2 (fibroblast); ADP,ATP carrier protein 2; ADP,ATP carrier protein, fibroblast isoform; ADP/ATP translocase 2; ANT 2; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5; Solute carrier family 25 member 5
Gene Aliases: 2F1; AAC2; ANT2; SLC25A5; T2; T3
UniProt ID: (Human) P05141
Entrez Gene ID: (Human) 292
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support