Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: SLC25A6, 5.
SLC25A5 and SLC25A6 are members of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this genes functions as a gated pore that translocates ADP from the cytoplasm into the mitochondrial matrix and ATP from the mitochondrial matrix into the cytoplasm. SLC25A5 forms a homodimer embedded in the inner mitochondria membrane. Suppressed expression of SLC25A5 gene has been shown to induce apoptosis and inhibit tumor growth. SLC25A6 is implicated in the function of the permability transition pore complex (PTPC), which regulates the release of mitochondrial products that induce apoptosis. The human genome contains several non-transcribed pseudogenes of this gene.[provided by RefSeq, Jun 2013].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenine nucleotide translocator 2; adenine nucleotide translocator 2 (fibroblast); Adenine nucleotide translocator 3; ADP,ATP carrier protein 2; ADP,ATP carrier protein 3; ADP,ATP carrier protein, fibroblast isoform; ADP,ATP carrier protein, isoform T2; ADP,ATP carrier protein, liver; ADP/ATP translocase 2; ADP/ATP translocase 3; ADP/ATP translocator of liver; ANT 2; ANT 3; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 6; Solute carrier family 25 member 5; Solute carrier family 25 member 6
Gene Aliases: 2F1; AAC2; AAC3; ANT; ANT 2; ANT 3; ANT2; ANT3; ANT3Y; CDABP0051; SLC25A5; SLC25A6; T2; T3
UniProt ID: (Human) P05141, (Human) P12236
Entrez Gene ID: (Human) 292, (Human) 293
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support