Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DFARTRLAAD VGKSGTEREF RGLGDCLVKI TKSDGIRGLY QGFSVSVQGI IIYRAAYFGV YDTAKGMLPD PKNTH
This gene is a member of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the mitochondrial matrix into the cytoplasm. The protein is implicated in the function of the permability transition pore complex (PTPC), which regulates the release of mitochondrial products that induce apoptosis. The human genome contains several non-transcribed pseudogenes of this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenine nucleotide translocator 3; ADP,ATP carrier protein 3; ADP,ATP carrier protein, isoform T2; ADP,ATP carrier protein, liver; ADP/ATP translocase 3; ADP/ATP translocator of liver; ANT 2; ANT 3; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 6; Solute carrier family 25 member 6
Gene Aliases: AAC3; ANT; ANT 2; ANT 3; ANT3; ANT3Y; CDABP0051; SLC25A6
UniProt ID: (Human) P12236
Entrez Gene ID: (Human) 293
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support