Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: RSLRVIVKEF QRIDVNVYFA SLQDYVIEKL EQCGFFDDNI RKDTFFLTVH DAILYLQNQV KSQEGQGSIL ETITLIQDCK D
Mutations in this gene are associated with Pendred syndrome, the most common form of syndromic deafness, an autosomal-recessive disease. It is highly homologous to the SLC26A3 gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pendrin; Sodium-independent chloride/iodide transporter; solute carrier family 26 (anion exchanger), member 4; Solute carrier family 26 member 4; truncated solute carrier family 26
Gene Aliases: DFNB4; EVA; PDS; SLC26A4; TDH2B
UniProt ID: (Human) O43511
Entrez Gene ID: (Human) 5172
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support