Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: RVFIKTIRRD IFGGLVLLKV KAKVRQCLQE RRTVPILFAS TVRRHPDKTA LIFEGTDTHW TFRQLDEYSS SVANFLQARG LASGDVAAIF MENRNEFVGL WLGMAKLGVE AALINTNLRR DALLHCLTTS RARALVFGSE MASAICEVHA SLDPSLSLFC SGSWEPGAVP PSTEHLDPLL KDAPKHLPSC PDKGF
This gene encodes a member of a family of fatty acid transport proteins, which are involved in translocation of long-chain fatty acids cross the plasma membrane. This protein is expressed at high levels on the apical side of mature enterocytes in the small intestine, and appears to be the principal fatty acid transporter in enterocytes. Clinical studies suggest this gene as a candidate gene for the insulin resistance syndrome. Mutations in this gene have been associated with ichthyosis prematurity syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACSVL4; Arachidonate--CoA ligase; FATP-4; fatty acid transport protein 4; Long-chain fatty acid transport protein 4; Long-chain-fatty-acid--CoA ligase; solute carrier family 27 (fatty acid transporter), member 4; Solute carrier family 27 member 4; Very long-chain acyl-CoA synthetase 4
Gene Aliases: ACSVL4; BB144259; FATP4; IPS; SLC27A4
UniProt ID: (Human) Q6P1M0, (Mouse) Q91VE0
Entrez Gene ID: (Human) 10999, (Mouse) 26569
Molecular Function:
transporter
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support