Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glucose transporter type 7; GLUT-7; solute carrier family 2, facilitated glucose transporter member 7
Gene Aliases: OTTMUSG00000010396
Entrez Gene ID: (Mouse) 435818, (Rat) 100362644
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support