Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
The protein encoded by the SLC30A2 gene is a zinc transporter that acts as a homodimer. The encoded protein plays a role in secreting zinc into breast milk. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Proton-coupled zinc antiporter SLC30A2; solute carrier family 30 (zinc transporter), member 2; Solute carrier family 30 member 2; Zinc transporter 2; ZnT-2
Gene Aliases: 6030430C21; PP12488; SLC30A2; TNZD; ZnT-2; ZNT2
UniProt ID: (Human) Q9BRI3, (Mouse) Q2HJ10
Entrez Gene ID: (Human) 7780, (Mouse) 230810
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support