Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MSVYSGKVLL QTTPPHVIGQ LDKLIREVST LDGVLEVRNE HFWTLGFGSL AGSVHVRIRR DANEQMVLAH VTNRLYTLVS TLTVQIFKDD WIRPALLSGP VAANVLNFSD HHVIPMPLLK GTDDLNPVTS TPAKPSSPPP EFSFNTPGKN VNPVILLNTQ TRPYGFGLNH GHTPYSSMLN QGLGVPGIGA TQGLRTGFTN IPSRYGTNNR IGQPRP
Zinc functions as a cofactor for numerous enzymes, nuclear factors, and hormones and as an intra- and intercellular signal ion. Members of the zinc transporter (ZNT)/SLC30 subfamily of the cation diffusion facilitator family, such as SLC30A6, permit cellular efflux of zinc (Seve et al., 2004).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: solute carrier family 30 (zinc transporter), member 6; Solute carrier family 30 member 6; Zinc transporter 6; ZnT-6
Gene Aliases: MST103; MSTP103; SLC30A6; ZNT6
UniProt ID: (Human) Q6NXT4
Entrez Gene ID: (Human) 55676
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support