Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Slc30A8 is a zinc efflux transporter involved in the accumulation of zinc in intracellular vesicles and is expressed at a high level in the pancreas, particularly in islets of Langerhans, and co-localizes with insulin in the secretory pathway granules of the insulin-secreting INS-1 cells. Allelic variants of Slc30A8 have been identified as major genetic risk factors for the development of Type 2 diabetes, but no genetic association has been found with Type 1 diabetes. Slc30A8 expression in b-cells was found to be influenced by cytokine expression, particularly IFN-gamma and IL-1 beta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Proton-coupled zinc antiporter SLC30A8; solute carrier family 30 (zinc transporter), member 8; Solute carrier family 30 member 8; Zinc transporter 8; zinc transporter ZnT-8; ZnT-8
Gene Aliases: SLC30A8; ZnT-8; ZNT8
UniProt ID: (Human) Q8IWU4
Entrez Gene ID: (Human) 169026
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support