Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: ALGQSFIFMT VVYFGPLTCS IITTTRKFFT ILASVILFAN PISPMQWVGT
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%
SLC35B1 belongs to the nucleotide-sugar transporter family and it is probable sugar transporter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP/ADP exchanger ER; AXER; Endoplasmic reticulum ATP/ADP translocase; hUGTrel1; Solute carrier family 35 member B1; solute carrier family 35, member B1; UDP-galactose transporter related; UDP-galactose transporter-related protein 1; UGTrel1
Gene Aliases: SLC35B1; UGTREL1
UniProt ID: (Human) P78383
Entrez Gene ID: (Human) 10237
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support