Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MDARWWAVVV LAAFPSLGAG GETPEAPPES WTQLWFFRFV VNAAGYASFM VPGYLLVQYF RRKNYLETGR GLCFPLVKAC VFGNEPKASD EVPLAPRTEA
Mediates the transport of adenosine 3'-phospho 5'-phosphosulfate (PAPS), from cytosol into Golgi. PAPS is a universal sulfuryl donor for sulfation events that take place in the Golgi. May indirectly participate in activation of the NF-kappa-B and MAPK pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-phosphoadenosine 5'-phosphosulfate transporter; Adenosine 3'-phospho 5'-phosphosulfate transporter 1; PAPS transporter 1; Putative MAPK-activating protein PM15; Putative NF-kappa-B-activating protein 48; solute carrier family 35 (adenosine 3'-phospho 5'-phosphosulfate transporter), member B2; Solute carrier family 35 member B2; solute carrier family 35 member B2 variant 2; solute carrier family 35, member B2
Gene Aliases: PAPST1; PSEC0149; SLC35B2; SLL; UGTrel4
UniProt ID: (Human) Q8TB61
Entrez Gene ID: (Human) 347734
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support